Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204466.1 | NP_001191395.1 | 20 | 2547 | ||
NM_001204467.1 | NP_001191396.1 | 21 | 2775 | |||
NM_001204468.1 | NP_001191397.1 | 21 | 2809 | |||
NM_005676.4 | NP_005667.2 | 21 | 2778 | |||
NM_152856.2 | NP_690595.1 | 20 | 2544 | |||
XM_005272677.4 | XP_005272734.1 | 21 | 2828 | |||
XM_005272678.4 | XP_005272735.1 | 20 | 2362 | |||
XM_005272679.4 | XP_005272736.1 | 20 | 2359 | |||
XM_006724563.1 | XP_006724626.1 | 17 | 1995 | |||
XM_011543989.2 | XP_011542291.1 | 13 | 1719 | |||
XM_017029884.1 | XP_016885373.1 | 20 | 2698 | |||
XM_017029885.1 | XP_016885374.1 | 20 | 2470 | |||
GenBank mRNA | AK024839.1 | - | 4 | 386 | ||
AK025854.1 | - | 9 | 1151 | |||
AK098115.1 | - | 2113 | ||||
AK225175.1 | - | 12 | 1450 | |||
AK292758.1 | - | 20 | 2498 | |||
AL137421.1 | - | 22 | 2667 | |||
BC003089.1 | - | 20 | 2483 | 3536058 | ||
BC004181.2 | - | 21 | 2731 | 2961094 | ||
BC008733.2 | - | 21 | 2763 | 3163064 | ||
BC024153.2 | - | 20 | 2393 | 3845258 | ||
BX537969.1 | - | 21 | 2760 | |||
D50912.1 | - | 21 | 2626 | |||
KU178411.1 | - | 19 | 2151 | |||
KU178412.1 | - | 20 | 2385 | |||
U35373.1 | - | 7 | 762 |