Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csgalnact1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107309.1 | NP_001100779.1 | 1 | 197 | ||
XM_006253026.2 | XP_006253088.1 | 2 | 653 | |||
XM_006253027.2 | XP_006253089.1 | 3 | 748 | |||
XM_006253028.2 | XP_006253090.1 | 3 | 1162 | |||
XM_006253029.2 | XP_006253091.1 | 3 | 968 | |||
XM_006253030.2 | XP_006253092.1 | 3 | 1052 | |||
XM_006253031.2 | XP_006253093.1 | 2 | 998 | |||
XM_008771110.1 | XP_008769332.1 | 3 | 782 | |||
XM_008771111.1 | XP_008769333.1 | 3 | 787 | |||
XM_008771112.1 | XP_008769334.1 | 3 | 788 | |||
XM_008771113.1 | XP_008769335.1 | 3 | 787 | |||
XM_008771114.1 | XP_008769336.1 | 3 | 786 | |||
XM_008771115.1 | XP_008769337.1 | 5 | 922 | |||
XM_008771116.1 | XP_008769338.1 | 3 | 926 | |||
GenBank mRNA | BC168857.1 | - | 1 | 421 | 7459880 |