Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csgalnact1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107309.1 | NP_001100779.1 | 7 | 1366 | ||
XM_006253026.2 | XP_006253088.1 | 8 | 1822 | |||
XM_006253027.2 | XP_006253089.1 | 9 | 1917 | |||
XM_006253028.2 | XP_006253090.1 | 9 | 2331 | |||
XM_006253029.2 | XP_006253091.1 | 9 | 2137 | |||
XM_006253030.2 | XP_006253092.1 | 9 | 2221 | |||
XM_006253031.2 | XP_006253093.1 | 8 | 2167 | |||
XM_008771110.1 | XP_008769332.1 | 9 | 1951 | |||
XM_008771111.1 | XP_008769333.1 | 9 | 1956 | |||
XM_008771112.1 | XP_008769334.1 | 9 | 1957 | |||
XM_008771113.1 | XP_008769335.1 | 9 | 1956 | |||
XM_008771114.1 | XP_008769336.1 | 9 | 1955 | |||
XM_008771115.1 | XP_008769337.1 | 11 | 2091 | |||
XM_008771116.1 | XP_008769338.1 | 9 | 2095 | |||
GenBank mRNA | BC168857.1 | - | 7 | 1590 | 7459880 |