Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107311.1 | NP_001100781.1 | 3 | 526 | ||
XM_006253156.2 | XP_006253218.1 | 3 | 544 | |||
XM_008771259.1 | XP_008769481.1 | 3 | 544 | |||
GenBank mRNA | BC158772.1 | - | 3 | 506 | 7128690 |