Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253168.1 | XP_006253230.1 | 7 | 977 | ||
XM_224863.6 | XP_224863.5 | 7 | 977 |