Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178093.2 | NP_835194.2 | 5 | 887 | ||
XM_006253180.2 | XP_006253242.1 | 9 | 3502 | |||
XM_006253181.2 | XP_006253243.1 | 9 | 3202 | |||
XM_006253182.2 | XP_006253244.1 | 7 | 1822 | |||
XM_008771280.1 | XP_008769502.1 | 6 | 1246 | |||
GenBank mRNA | AB190507.1 | - | 5 | 964 | ||
AY208915.1 | - | 5 | 655 | |||
AY208916.1 | - | 4 | 373 | |||
BC072537.1 | - | 5 | 887 | |||
BC166591.1 | - | 5 | 936 | 7938925 | ||
EU417559.1 | - | 8 | 3005 | |||
GU245886.1 | - | 7 | 895 |