Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256120.1 | NP_001243049.1 | 6, 7 | 618 | ||
NM_003491.3 | NP_003482.1 | 6, 7 | 636 | |||
GenBank mRNA | AY183134.1 | - | 5, 6 | 697 | ||
BC000308.2 | - | 6, 7 | 497 | 2820955 | ||
BC019312.1 | - | 6, 7 | 497 | 2820955 | ||
BI192945.1 | - | 6, 7 | 433 | |||
CR979270.1 | - | 6, 7 | 636 | |||
X77588.1 | - | 6, 7 | 481 |