Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007598.3 | NP_001007599.3 | 2 | 50 | ||
GenBank mRNA | BC060589.1 | - | 2 | 54 | 6888030 | |
BC086561.1 | - | 1 | 32 | 7304371 | ||
FQ210235.1 | - | 2 | 46 | |||
FQ211371.1 | - | 2 | 50 | |||
FQ212163.1 | - | 2 | 49 | |||
FQ212174.1 | - | 2 | 50 | |||
FQ212384.1 | - | 2 | 54 | |||
FQ212431.1 | - | 2 | 49 | |||
FQ216865.1 | - | 2 | 50 | |||
FQ220825.1 | - | 2 | 48 | |||
FQ221003.1 | - | 2 | 50 | |||
FQ221028.1 | - | 2 | 50 | |||
FQ221603.1 | - | 2 | 50 | |||
FQ221794.1 | - | 2 | 52 | |||
FQ222130.1 | - | 2 | 52 | |||
FQ222146.1 | - | 2 | 50 | |||
FQ222204.1 | - | 2 | 50 | |||
FQ222278.1 | - | 2 | 48 | |||
FQ222508.1 | - | 2 | 52 | |||
FQ222509.1 | - | 2 | 50 | |||
FQ222630.1 | - | 2 | 49 | |||
FQ222743.1 | - | 2 | 50 | |||
FQ223008.1 | - | 2 | 64 | |||
FQ223059.1 | - | 2 | 50 | |||
FQ223395.1 | - | 2 | 52 | |||
FQ223420.1 | - | 2 | 48 | |||
FQ223651.1 | - | 2 | 45 | |||
FQ224555.1 | - | 2 | 50 | |||
FQ228320.1 | - | 2 | 51 | |||
FQ228532.1 | - | 2 | 62 | |||
FQ229128.1 | - | 2 | 48 | |||
X51706.1 | - | 1 | 32 |
Gene Symbol: LOC100360449 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729147.2 | XP_002729193.1 | 1 | 56 |
Gene Symbol: LOC100364457 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748774.3 | XP_003748822.1 | 1 | 63 |