Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107321.1 | NP_001100791.1 | 2, 3 | 721 | ||
XM_008771355.1 | XP_008769577.1 | 2, 3 | 3864 |