Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014235.4 | NP_055050.1 | 3 | 254 | ||
GenBank mRNA | AK000405.1 | - | 3 | 189 | ||
AK293227.1 | - | 2 | 416 | |||
BC043346.2 | - | 3 | 181 | 6058941 | ||
BC053589.1 | - | 3 | 215 | 5246329 | ||
DR003955.1 | - | 3 | 194 |