Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014005.1 | NP_001014027.1 | 1 | 345 | ||
XM_006243774.2 | XP_006243836.1 | 1 | 545 | |||
XM_008766514.1 | XP_008764736.1 | 2 | 427 | |||
GenBank mRNA | BC083731.1 | - | 1 | 345 | 7191343 | |
FQ229045.1 | - | 1 | 416 |