Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014005.1 | NP_001014027.1 | 3 | 1357 | ||
XM_006243774.2 | XP_006243836.1 | 3 | 1605 | |||
XM_008766514.1 | XP_008764736.1 | 3 | 1384 | |||
GenBank mRNA | BC083731.1 | - | 3 | 1357 | 7191343 |