Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039505.2 | NP_001034594.1 | 3 | 1462 | ||
XM_008758689.1 | XP_008756911.1 | 3 | 1519 | |||
GenBank mRNA | AW434973.1 | - | 1 | 126 |