Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254738.1 | NP_001241667.1 | 1 | 346 | ||
NM_005168.4 | NP_005159.1 | 2 | 253 | |||
GenBank mRNA | AF498969.1 | - | 1 | 68 | ||
AK298007.1 | - | 2 | 223 | |||
AK313224.1 | - | 1 | 320 | |||
AL702945.1 | - | 1 | 319 | |||
BC012513.1 | - | 2 | 208 | 4453777 | ||
BQ440721.1 | - | 1 | 322 | |||
BT006769.1 | - | 1 | 68 | |||
CR542038.1 | - | 1 | 23 | |||
CR542070.1 | - | 1 | 23 | |||
DC367501.1 | - | 2 | 254 | |||
S82240.1 | - | 1 | 47 | |||
X95282.1 | - | 1 | 68 | |||
X97758.1 | - | 1 | 290 |