Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013137.2 | NP_001013155.1 | 2, 3 | 364 | ||
GenBank mRNA | BC101896.1 | - | 2, 3 | 364 | 7932648 |