Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047095.1 | NP_001040560.1 | 5 | 522 | ||
GenBank mRNA | AB256043.1 | - | 5 | 522 | ||
AM258958.1 | - | 5 | 522 | |||
BC158562.1 | - | 5 | 526 | 9088902 |