Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nutm2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771552.1 | XP_008769774.1 | 2 | 2623 | ||
XM_225188.7 | XP_225188.5 | 3 | 976 |