Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100572.1 | NP_001094042.1 | 6 | 795 | ||
XM_006253706.2 | XP_006253768.1 | 6 | 685 | |||
XM_008771533.1 | XP_008769755.1 | 7 | 893 |
Gene Symbol: LOC103690033 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771517.1 | XP_008769739.1 | 841 | |||
XM_008771518.1 | XP_008769740.1 | 899 | ||||
XM_008771519.1 | XP_008769741.1 | 685 |