Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014008.1 | NP_001014030.1 | 810 | |||
XM_006253695.2 | XP_006253757.1 | 2680 | ||||
GenBank mRNA | BC083711.1 | - | 810 | 7190620 |
Gene Symbol: NEWGENE_1549776 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253707.2 | XP_006253769.1 | 6 | 2680 | ||
XM_008771557.1 | XP_008769779.1 | 5 | 2756 |