Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004255.1 | NP_001004255.1 | 7 | 737 | ||
XM_006253756.2 | XP_006253818.1 | 7 | 760 | |||
GenBank mRNA | BC079344.1 | - | 7 | 737 | 7107191 |