Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bloc1s5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107347.1 | NP_001100817.1 | 2 | 242 | ||
XM_006253807.2 | XP_006253869.1 | 2 | 387 | |||
GenBank mRNA | BC166529.1 | - | 2 | 168 | 7189489 |