Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mylk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253874.2 | XP_006253936.1 | 9 | 1827 |
Gene Symbol: LOC103689985 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253865.2 | XP_006253927.1 | 8 | 1564 | ||
XM_008771601.1 | XP_008769823.1 | 8 | 1564 |