Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201561.1 | NP_963855.1 | 4 | 309 | ||
XM_008771653.1 | XP_008769875.1 | 5 | 715 | |||
GenBank mRNA | AY513659.1 | - | 4 | 309 |