Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006015.4 | NP_006006.3 | 20 | 7166 | ||
NM_139135.2 | NP_624361.1 | 20 | 6515 | |||
GenBank mRNA | AF217506.1 | - | 1 | 400 | ||
AF219114.1 | - | 20 | 5755 | |||
AF231056.1 | - | 20 | 7171 | |||
AF265208.1 | - | 20 | 5721 | |||
AF268913.1 | - | 19 | 4993 | |||
AF521670.1 | - | 20 | 5935 | |||
AK027442.1 | - | 1 | 1670 | |||
AK027467.1 | - | 1 | 1504 | |||
AK027655.1 | - | 6 | 2210 | |||
AK074940.1 | - | 3 | 2225 | |||
AK223275.1 | - | 9 | 3483 | |||
AK308363.1 | - | 4 | 2289 | |||
BC112895.1 | - | 1 | 432 |