Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270417.1 | NP_001257346.1 | 8 | 1335 | ||
GenBank mRNA | AB574169.1 | - | 8 | 1335 |
Gene Symbol: LOC100909597 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764495.1 | XP_008762717.1 | 3 | 1208 |