Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Socs6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271149.1 | NP_001258078.1 | 2 | 1542 | ||
XM_006255033.2 | XP_006255095.1 | 3 | 1895 | |||
XM_006255034.2 | XP_006255096.1 | 3 | 1779 | |||
XM_008772132.1 | XP_008770354.1 | 2 | 1790 |