Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Malt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751798.3 | XP_003751846.1 | 10 | 1351 | ||
XM_006254888.2 | XP_006254950.1 | 10 | 1351 | |||
XM_225927.8 | XP_225927.5 | 9 | 1317 |