Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ammecr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107399.1 | NP_001100869.1 | 7 | 1053 | ||
XM_006254590.2 | XP_006254652.1 | 7 | 1867 | |||
XM_006254591.2 | XP_006254653.1 | 7 | 1904 | |||
XM_006254593.2 | XP_006254655.1 | 7 | 1304 |