Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004656.3 | NP_004647.1 | 4 | 441 | ||
XM_011534149.2 | XP_011532451.1 | 4 | 437 | |||
XM_011534150.2 | XP_011532452.1 | 4 | 437 | |||
XM_011534151.2 | XP_011532453.1 | 4 | 437 | |||
XM_011534152.1 | XP_011532454.1 | 4 | 437 | |||
XM_017007303.1 | XP_016862792.1 | 4 | 437 | |||
GenBank mRNA | AF045581.1 | - | 4 | 253 | ||
AK292608.1 | - | 4 | 375 | |||
BC001596.1 | - | 4 | 329 | 3543914 | ||
D87462.2 | - | 4 | 338 | |||
D88812.1 | - | 4 | 214 |