Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006768.4 | NP_006759.3 | 9 | 1328 | ||
XM_005253944.4 | XP_005254001.1 | 9 | 1428 | |||
XM_011538789.2 | XP_011537091.1 | 8 | 1098 | |||
XM_017019992.1 | XP_016875481.1 | 8 | 1136 | |||
XM_017019993.1 | XP_016875482.1 | 8 | 1119 | |||
XM_017019994.1 | XP_016875483.1 | 7 | 994 | |||
XM_017019995.1 | XP_016875484.1 | 7 | 759 | |||
GenBank mRNA | AB208878.1 | - | 9 | 1283 | ||
AF035620.1 | - | 9 | 1263 | |||
AF035950.1 | - | 2 | 59 | |||
AK290379.1 | - | 9 | 1268 | |||
AK299326.1 | - | 6 | 634 | |||
AK308279.1 | - | 9 | 1175 | |||
AY332222.1 | - | 9 | 1263 | |||
BC136698.1 | - | 9 | 1263 | 9020688 | ||
BC136699.1 | - | 9 | 1263 | 9020689 |