Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006768.4 | NP_006759.3 | 9, 10 | 1419 | ||
XM_005253944.4 | XP_005254001.1 | 9, 10 | 1519 | |||
XM_011538789.2 | XP_011537091.1 | 8, 9 | 1189 | |||
XM_017019992.1 | XP_016875481.1 | 8, 9 | 1227 | |||
XM_017019993.1 | XP_016875482.1 | 8, 9 | 1210 | |||
XM_017019994.1 | XP_016875483.1 | 7, 8 | 1085 | |||
XM_017019995.1 | XP_016875484.1 | 7, 8 | 850 | |||
GenBank mRNA | AB208878.1 | - | 9, 10 | 1374 | ||
AF035620.1 | - | 9, 10 | 1354 | |||
AF035950.1 | - | 2, 3 | 150 | |||
AK290379.1 | - | 9, 10 | 1359 | |||
AK299326.1 | - | 6, 7 | 725 | |||
AK308279.1 | - | 9, 10 | 1266 | |||
AY332222.1 | - | 9, 10 | 1354 | |||
BC136698.1 | - | 9, 10 | 1354 | 9020688 | ||
BC136699.1 | - | 9, 10 | 1354 | 9020689 |