Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013142.1 | NP_001013160.1 | 2 | 371 | ||
GenBank mRNA | BC078975.1 | - | 2 | 371 | 7111936 |