Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfap20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037978.1 | NP_001033067.1 | 1, 2 | 252 | ||
XM_006255083.1 | XP_006255145.1 | 1, 2 | 338 | |||
GenBank mRNA | BC099768.1 | - | 1, 2 | 252 | 7932140 |