Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kifc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103352.1 | NP_001096822.1 | 14 | 2349 | ||
XM_006255086.2 | XP_006255148.1 | 18 | 2651 | |||
XM_006255087.2 | XP_006255149.1 | 18 | 2651 | |||
XM_006255088.2 | XP_006255150.1 | 18 | 2651 | |||
XM_008772286.1 | XP_008770508.1 | 18 | 2648 | |||
XM_008772287.1 | XP_008770509.1 | 16 | 2330 | |||
XM_008772288.1 | XP_008770510.1 | 18 | 2641 | |||
XM_008772289.1 | XP_008770511.1 | 17 | 2424 | |||
XM_008772290.1 | XP_008770512.1 | 16 | 2513 | |||
XM_008772291.1 | XP_008770513.1 | 17 | 2491 | |||
XM_008772292.1 | XP_008770514.1 | 15 | 2120 | |||
XM_008772293.1 | XP_008770515.1 | 14 | 2127 | |||
XM_008772294.1 | XP_008770516.1 | 15 | 4286 | |||
XM_008772295.1 | XP_008770517.1 | 16 | 2319 | |||
XM_008772296.1 | XP_008770518.1 | 14 | 4142 | |||
GenBank mRNA | BC128748.1 | - | 14 | 2349 | 7099524 |