Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107410.1 | NP_001100880.1 | 6, 7 | 608 | ||
XM_006255101.2 | XP_006255163.1 | 7, 8 | 678 | |||
XM_008772297.1 | XP_008770519.1 | 7, 8 | 666 | |||
XM_008772298.1 | XP_008770520.1 | 7, 8 | 664 | |||
XM_008772299.1 | XP_008770521.1 | 7, 8 | 666 | |||
XM_008772300.1 | XP_008770522.1 | 6, 7 | 774 | |||
XM_008772301.1 | XP_008770523.1 | 7, 8 | 702 | |||
XM_008772302.1 | XP_008770524.1 | 7, 8 | 624 | |||
XM_008772303.1 | XP_008770525.1 | 7, 8 | 656 | |||
XM_008772304.1 | XP_008770526.1 | 7, 8 | 963 | |||
XM_008772305.1 | XP_008770527.1 | 7, 8 | 624 | |||
XM_008772306.1 | XP_008770528.1 | 7, 8 | 665 | |||
XM_008772307.1 | XP_008770529.1 | 7, 8 | 665 | |||
XM_008772308.1 | XP_008770530.1 | 7, 8 | 697 | |||
XM_008772309.1 | XP_008770531.1 | 7, 8 | 673 | |||
XM_008772310.1 | XP_008770532.1 | 7, 8 | 651 | |||
XM_008772311.1 | XP_008770533.1 | 7, 8 | 616 |
Set Membership: |
G-protein coupled receptor |