Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178010.2 | NP_001171481.1 | 8 | 811 | ||
NM_001178011.2 | NP_001171482.1 | 6 | 577 | |||
NM_003504.4 | NP_003495.1 | 7 | 715 | |||
XM_005261285.2 | XP_005261342.1 | 9 | 904 | |||
XM_005261286.3 | XP_005261343.1 | 8 | 808 | |||
XM_011530415.1 | XP_011528717.1 | 8 | 808 | |||
XM_011530416.1 | XP_011528718.1 | 7 | 611 | |||
XM_011530417.2 | XP_011528719.1 | 7 | 871 | |||
XM_011530418.2 | XP_011528720.1 | 7 | 871 | |||
XM_017028966.1 | XP_016884455.1 | 2 | 190 | |||
GenBank mRNA | AF053074.1 | - | 7 | 596 | ||
AF062495.1 | - | 7 | 588 | |||
AF081535.1 | - | 7 | 603 | |||
AJ223728.1 | - | 7 | 602 | |||
AK293123.1 | - | 6 | 512 | |||
AK293338.1 | - | 8 | 708 | |||
AK302590.1 | - | 8 | 699 | |||
AK303274.1 | - | 7 | 635 | |||
AY358971.1 | - | 7 | 638 | |||
BC006232.2 | - | 7 | 633 | 3946493 | ||
BC010022.2 | - | 7 | 618 | 2964592 | ||
BT006792.1 | - | 7 | 572 | |||
CT841513.1 | - | 7 | 576 |