Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpgrip1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107414.1 | NP_001100884.1 | 8 | 1019 | ||
XM_008772313.1 | XP_008770535.1 | 7 | 1447 |