Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107419.1 | NP_001100889.1 | 10, 11 | 1145 | ||
XM_006255401.1 | XP_006255463.1 | 11, 12 | 1936 |