Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107421.1 | NP_001100891.1 | 3 | 1157 | ||
XM_008772497.1 | XP_008770719.1 | 3 | 2211 | |||
XM_008772498.1 | XP_008770720.1 | 3 | 1751 | |||
GenBank mRNA | BC161901.1 | - | 3 | 1139 | 8367133 |