Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtss1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191558.1 | NP_001178487.1 | 14 | 1664 | ||
XM_008772512.1 | XP_008770734.1 | 15 | 1775 | |||
XM_008772513.1 | XP_008770735.1 | 14 | 1725 |