Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtss1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772512.1 | XP_008770734.1 | 11 | 903 | ||
XM_008772513.1 | XP_008770735.1 | 10 | 853 |