Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gse1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255748.2 | XP_006255810.1 | 9, 10 | 2504 | ||
XM_008772656.1 | XP_008770878.1 | 8, 9 | 8564 | |||
XM_008772657.1 | XP_008770879.1 | 8, 9 | 8564 | |||
XM_008772658.1 | XP_008770880.1 | 10, 11 | 2474 | |||
XM_008772659.1 | XP_008770881.1 | 9, 10 | 3832 | |||
XM_008772660.1 | XP_008770882.1 | 8, 9 | 2285 | |||
XM_008772661.1 | XP_008770883.1 | 8, 9 | 2879 | |||
XM_008772662.1 | XP_008770884.1 | 8, 9 | 3612 | |||
XM_226533.8 | XP_226533.5 | 9, 10 | 3098 |