Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728603.4 | XP_002728649.1 | 11 | 2096 | ||
GenBank mRNA | BC079416.1 | - | 11 | 1455 |