Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135715.1 | NP_001129187.1 | 9 | 2687 | ||
XM_006255815.2 | XP_006255877.1 | 9 | 2978 |