Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssx2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175597.3 | NP_783187.1 | 14 | 1890 | ||
XM_006233454.2 | XP_006233516.1 | 13 | 1703 | |||
XM_006233455.2 | XP_006233517.1 | 13 | 1852 | |||
XM_006233458.2 | XP_006233520.1 | 10 | 1833 | |||
XM_008761498.1 | XP_008759720.1 | 12 | 1758 | |||
GenBank mRNA | AF532970.1 | - | 12 | 1603 | ||
BC078687.1 | - | 14 | 1890 | 7115887 |