Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038615.1 | NP_001033704.1 | 15, 16 | 4690 | ||
XM_006227871.2 | XP_006227933.1 | 18, 19 | 5195 | |||
XM_008758732.1 | XP_008756954.1 | 17, 18 | 5006 | |||
XM_008758733.1 | XP_008756955.1 | 15, 16 | 4844 | |||
GenBank mRNA | DQ256268.1 | - | 15, 16 | 4690 |