Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107460.1 | NP_001100930.1 | 1 | 161 | ||
XM_008758705.1 | XP_008756927.1 | 1 | 160 |