Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcp10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107461.1 | NP_001100931.1 | 9 | 1342 | ||
XM_006227927.2 | XP_006227989.1 | 13 | 1651 | |||
XM_006227928.2 | XP_006227990.1 | 12 | 1549 | |||
XM_006227930.2 | XP_006227992.1 | 11 | 1408 | |||
XM_008758757.1 | XP_008756979.1 | 10 | 1443 | |||
XM_008758758.1 | XP_008756980.1 | 9 | 1302 | |||
XM_008758759.1 | XP_008756981.1 | 12 | 1588 | |||
GenBank mRNA | BC166440.1 | - | 11 | 1339 | 7455355 |