Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps37c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107463.1 | NP_001100933.1 | 3 | 397 | ||
XM_006231033.2 | XP_006231095.1 | 3 | 455 | |||
XM_006231034.2 | XP_006231096.1 | 3 | 459 | |||
XM_006231035.2 | XP_006231097.1 | 3 | 1421 | |||
GenBank mRNA | BC169040.1 | - | 3 | 390 | 9024199 |
Gene Symbol: LOC100910929 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749059.3 | XP_003749107.1 | 3 | 455 | ||
XM_006231078.2 | XP_006231140.1 | 3 | 459 | |||
XM_006231079.2 | XP_006231141.1 | 3 | 1421 |