Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp772 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107472.1 | NP_001100942.1 | 5 | 1317 | ||
GenBank mRNA | BC088175.1 | - | 5 | 1249 | ||
BC107472.1 | - | 5 | 1245 |